ENA341293227 Enamine

3-fluoro-4-(1H-1,2,4-triazol-1-yl)benzoic acid

  • MDL number MFCD11156563
  • Cas Number : 1020934-33-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 285...287
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88731

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-88731
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart