ENA341293229 Enamine

1-(2-bromophenyl)butan-2-one

  • MDL number MFCD12153368
  • Cas Number : 1179882-16-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88734

Safety Information

msds https://www.enaminestore.com/catalog/EN300-88734
Availability details for
Quantity
  • Availability
  • Add to Cart