ENA341293233 Enamine

6-chloro-N1-(2-methoxyethyl)benzene-1,2-diamine

  • MDL number MFCD11186910
  • Cas Number : 1040083-79-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88745

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-88745
Availability details for
Quantity
  • Availability
  • Add to Cart