ENA341293242 Enamine

1-(3-methyl-1H-1,2,4-triazol-5-yl)propan-1-amine dihydrochloride

  • MDL number MFCD20441622
  • Cas Number : 1354960-05-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-88755

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-88755
Availability details for
Quantity
  • Availability
  • Add to Cart