ENA341293248 Enamine

4-(4-methylphenyl)-1,2,5-oxadiazol-3-amine

  • MDL number MFCD02647894
  • Cas Number : 99817-28-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-88762

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-88762
Availability details for
Quantity
  • Availability
  • Add to Cart