ENA341293249 Enamine

4-(chloromethyl)-2-(methanesulfonylmethyl)-1,3-thiazole

  • MDL number MFCD16766330
  • Cas Number : 1249560-17-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88765

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-88765
Availability details for
Quantity
  • Availability
  • Add to Cart