ENA341293252 Enamine

3-methyl-2-[(2,2,2-trifluoroethyl)amino]butanoic acid

  • MDL number MFCD12176650
  • Molecular Weight : 199.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88770

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-88770
Availability details for
Quantity
  • Availability
  • Add to Cart