ENA341293286 Enamine

2-(trifluoromethoxy)ethanesulfonyl chloride

  • MDL number MFCD11858188
  • Cas Number : 1118786-91-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88807

Safety Information

msds https://www.enaminestore.com/catalog/EN300-88807
Availability details for
Quantity
  • Availability
  • Add to Cart