ENA341293297 Enamine

3-({[4-(propan-2-yl)benzene]sulfonyl}methyl)furan-2-carboxylic acid

  • MDL number MFCD13467233
  • Cas Number : 1094269-20-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 156...158
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88824

Safety Information

msds https://www.enaminestore.com/catalog/EN300-88824
Availability details for
Quantity
  • Availability
  • Add to Cart