ENA341293301 Enamine

2-(chloromethyl)-5-(2-fluorophenyl)-1,3-oxazole

  • MDL number MFCD09835625
  • Cas Number : 1152568-80-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88829

Safety Information

msds https://www.enaminestore.com/catalog/EN300-88829
Availability details for
Quantity
  • Availability
  • Add to Cart