ENA341293334 Enamine

(2-morpholin-4-ylpyrimidin-5-yl)methylamine

  • MDL number MFCD10696914
  • Cas Number : 944899-76-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 42...44
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88863

Safety Information

msds https://www.enaminestore.com/catalog/EN300-88863
Availability details for
Quantity
  • Availability
  • Add to Cart