ENA341293408 Enamine

1,1-dioxo-2H,3H,5H-1lambda6-thieno[2,3-c]pyrrole-4-sulfonyl chloride

  • MDL number MFCD12913290
  • Cas Number : 1210918-13-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88939

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-88939
Availability details for
Quantity
  • Availability
  • Add to Cart