ENA341293438 Enamine

2-(6-hydroxy-2-oxo-2H-chromen-4-yl)acetic acid

  • MDL number MFCD13196342
  • Cas Number : 68747-26-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88969

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-88969
Availability details for
Quantity
  • Availability
  • Add to Cart