ENA341293441 Enamine

6-chloro-2,4'-bipyridine-5-carbonitrile

  • MDL number MFCD00137929
  • Cas Number : 63219-03-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88973

Safety Information

msds https://www.enaminestore.com/catalog/EN300-88973
Availability details for
Quantity
  • Availability
  • Add to Cart