ENA341293445 Enamine

1,3,7-triazaspiro[4.5]decane-2,4-dione

  • MDL number MFCD07776799
  • Cas Number : 78222-09-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88977

Safety Information

msds https://www.enaminestore.com/catalog/EN300-88977
Availability details for
Quantity
  • Availability
  • Add to Cart