ENA341293538 Enamine

N-(4-aminocyclohexyl)piperidine-4-carboxamide dihydrochloride

  • MDL number MFCD20501928
  • Cas Number : 1354950-03-0
  • Molecular Weight : 298.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-89854

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-89854
Availability details for
Quantity
  • Availability
  • Add to Cart