ENA341293571 Enamine

7-chloro-1H-1,3-benzodiazole

  • MDL number MFCD11694327
  • Cas Number : 16931-35-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-89118

Safety Information

msds https://www.enaminestore.com/catalog/EN300-89118
Availability details for
Quantity
  • Availability
  • Add to Cart