ENA341293601 Enamine

2-amino-5-iodophenol

  • MDL number MFCD14705905
  • Cas Number : 99968-80-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 104...106
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-89148

Safety Information

msds https://www.enaminestore.com/catalog/EN300-89148
Availability details for
Quantity
  • Availability
  • Add to Cart