ENA341293619 Enamine

1-(4-methanesulfonyl-3-methoxyphenyl)ethan-1-amine

  • MDL number MFCD14705921
  • Cas Number : 1235439-70-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-89167

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-89167
Availability details for
Quantity
  • Availability
  • Add to Cart