ENA341293621 Enamine

2-[4-(trifluoromethyl)-1,3-thiazol-2-yl]ethan-1-amine

  • MDL number MFCD05221347
  • Cas Number : 642081-23-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-89169

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-89169
Availability details for
Quantity
  • Availability
  • Add to Cart