ENA341293675 Enamine

(1R)-1-(adamantan-1-yl)ethan-1-ol

  • Cas Number : 91410-69-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-89224

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-89224
Availability details for
Quantity
  • Availability
  • Add to Cart