ENA341293677 Enamine

(1S)-1-(adamantan-1-yl)ethan-1-amine

  • Cas Number : 887336-06-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-89226

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-89226
Availability details for
Quantity
  • Availability
  • Add to Cart