ENA341293691 Enamine

(1S)-1-[4-(methylsulfanyl)phenyl]ethan-1-ol

  • MDL number MFCD14705948
  • Cas Number : 179613-06-0
  • Molecular Weight : 168.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-89240

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-89240
Availability details for
Quantity
  • Availability
  • Add to Cart