ENA341293700 Enamine

3-[(1S)-1-aminoethyl]phenol

  • MDL number MFCD06761830
  • Cas Number : 123982-81-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-89249

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-89249
Availability details for
Quantity
  • Availability
  • Add to Cart