ENA341293736 Enamine

(1S)-1-(5-bromothiophen-2-yl)ethan-1-ol

  • MDL number MFCD14705975
  • Cas Number : 222609-65-6
  • Molecular Weight : 207.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-89285

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-89285
Availability details for
Quantity
  • Availability
  • Add to Cart