ENA341293764 Enamine

(1S)-5-chloro-2,3-dihydro-1H-inden-1-ol

  • MDL number MFCD14705987
  • Cas Number : 1380394-45-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 90...92
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-89313

Safety Information

msds https://www.enaminestore.com/catalog/EN300-89313
Availability details for
Quantity
  • Availability
  • Add to Cart