ENA341293803 Enamine

4-chloro-6-methyl-2,2'-bipyrimidine

  • MDL number MFCD12093754
  • Cas Number : 1153093-46-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-89352

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-89352
Availability details for
Quantity
  • Availability
  • Add to Cart