ENA341293824 Enamine

4-(aminomethyl)-N,N-dimethyl-1,3-thiazol-2-amine

  • MDL number MFCD09880808
  • Cas Number : 1023818-77-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-89375

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-89375
Availability details for
Quantity
  • Availability
  • Add to Cart