ENA341293911 Enamine

4-[(4-methyl-1,3-thiazol-2-yl)amino]benzoic acid

  • MDL number MFCD02946200
  • Cas Number : 33942-54-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-89468

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-89468
Availability details for
Quantity
  • Availability
  • Add to Cart