ENA341293919 Enamine

4-amino-2-iodobenzoic acid

  • MDL number MFCD16547805
  • Cas Number : 73655-51-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160...162
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-89476

Safety Information

msds https://www.enaminestore.com/catalog/EN300-89476
Availability details for
Quantity
  • Availability
  • Add to Cart