ENA341293957 Enamine

1,2,3,4-tetrahydroquinoline-5-sulfonamide

  • MDL number MFCD12174259
  • Cas Number : 1155515-51-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-89514

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-89514
Availability details for
Quantity
  • Availability
  • Add to Cart