ENA341293996 Enamine

2-chloro-4-methyl-5-nitropyrimidine

  • MDL number MFCD16990715
  • Cas Number : 97821-70-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-89553

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-89553
Availability details for
Quantity
  • Availability
  • Add to Cart