ENA341294016 Enamine

5-(4-fluorophenoxy)-1,2,3,4-tetrahydroisoquinolin-8-amine

  • MDL number MFCD16990731
  • Cas Number : 1251924-58-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-89573

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-89573
Availability details for
Quantity
  • Availability
  • Add to Cart