ENA341294167 Enamine

1-amino-3-phenylcyclobutane-1-carboxylic acid

  • MDL number MFCD17480495
  • Cas Number : 17382-09-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-89725

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-89725
Availability details for
Quantity
  • Availability
  • Add to Cart