ENA341294250 Enamine

methyl 4-oxo-2,3,4,5-tetrahydro-1H-1,5-benzodiazepine-7-carboxylate

  • MDL number MFCD17977182
  • Cas Number : 465542-64-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-89808

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-89808
Availability details for
Quantity
  • Availability
  • Add to Cart