ENA341294270 Enamine

1,1,1-trifluoro-2-(piperidin-4-yl)propan-2-ol

  • MDL number MFCD17977193
  • Cas Number : 1227068-37-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 122...124
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-89829

Safety Information

msds https://www.enaminestore.com/catalog/EN300-89829
Availability details for
Quantity
  • Availability
  • Add to Cart