ENA341294275 Enamine

2,2,2-trifluoro-1-(piperidin-2-yl)ethan-1-ol

  • MDL number MFCD17977198
  • Cas Number : 1258651-25-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 94...96
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-89834

Safety Information

msds https://www.enaminestore.com/catalog/EN300-89834
Availability details for
Quantity
  • Availability
  • Add to Cart