ENA341294298 Enamine

3-cyanonaphthalene-1-carboxylic acid

  • MDL number MFCD17977212
  • Cas Number : 3839-18-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-89858

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-89858
Availability details for
Quantity
  • Availability
  • Add to Cart