ENA341294314 Enamine

6-ethoxypyridin-2-amine

  • MDL number MFCD00235129
  • Cas Number : 768-42-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 238...240
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-89876

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-89876
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart