ENA341294318 Enamine

4-(1,3-thiazol-4-yl)phenol

  • MDL number MFCD17977224
  • Cas Number : 68535-57-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-89880

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-89880
Availability details for
Quantity
  • Availability
  • Add to Cart