ENA341294343 Enamine

8-azabicyclo[5.2.0]nonane

  • MDL number MFCD17977242
  • Cas Number : 39872-95-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-89907

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-89907
Availability details for
Quantity
  • Availability
  • Add to Cart