ENA341294348 Enamine

4-[(pyrrolidin-3-yl)methyl]morpholine

  • MDL number MFCD09036034
  • Cas Number : 933704-68-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-89912

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-89912
Availability details for
Quantity
  • Availability
  • Add to Cart