ENA341294416 Enamine

2-{3,5-dioxo-10-oxa-4-azatricyclo[5.2.1.0^{2,6}]decan-4-yl}propanoic acid

  • MDL number MFCD20501930
  • Cas Number : 154902-09-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 189...191
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-89982

Safety Information

msds https://www.enaminestore.com/catalog/EN300-89982
Availability details for
Quantity
  • Availability
  • Add to Cart