ENA341294487 Enamine

6-(chloromethyl)-N,N-dimethylpyridin-2-amine

  • MDL number MFCD10697582
  • Cas Number : 405103-62-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90057

Safety Information

msds https://www.enaminestore.com/catalog/EN300-90057
Availability details for
Quantity
  • Availability
  • Add to Cart