ENA341294505 Enamine

8-aminoquinoline-6-carboxamide

  • MDL number MFCD18089647
  • Cas Number : 1308644-45-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-90079

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-90079
Availability details for
Quantity
  • Availability
  • Add to Cart