ENA341294525 Enamine

methyl 3-(piperidin-4-yl)-1H-indole-5-carboxylate

  • MDL number MFCD18089664
  • Cas Number : 173150-62-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90100

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-90100
Availability details for
Quantity
  • Availability
  • Add to Cart