ENA341294526 Enamine

3-(piperidin-4-yl)-1H-indole-5-carboxamide

  • MDL number MFCD18089665
  • Cas Number : 177940-51-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90101

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-90101
Availability details for
Quantity
  • Availability
  • Add to Cart