ENA341294607 Enamine

1-(5-bromo-2-methylthiophen-3-yl)ethan-1-one

  • MDL number MFCD20501932
  • Cas Number : 1001671-82-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90164

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-90164
Availability details for
Quantity
  • Availability
  • Add to Cart