ENA341294610 Enamine

4-methanesulfinylbutan-2-amine hydrochloride

  • MDL number MFCD20501935
  • Cas Number : 1354950-26-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90393

Safety Information

msds https://www.enaminestore.com/catalog/EN300-90393
Availability details for
Quantity
  • Availability
  • Add to Cart