ENA341294612 Enamine

5-(2-aminoethyl)-1,2,4-oxadiazol-3-amine hydrochloride

  • MDL number MFCD20501937
  • Cas Number : 1354960-93-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-90482

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-90482
Availability details for
Quantity
  • Availability
  • Add to Cart